accessibility menu, dialog, popup

UserName

CAS RN 181183-52-8

H3CNCH3HNSOONOHHOOOHO

CAS RN 181183-52-8

IUPAC Name: 2-hydroxybutanedioic acid; dimethyl(2-{5-[(pyrrolidine-1-sulfonyl)methyl]-1H-indol-3-yl}ethyl)amine
Synonyms: almotriptan malate
Molecular Weight (g/mol): 469.55
Molecular Formula: C21H31N3O7S
InChi Key: QHATUKWEVNMHRY-UHFFFAOYNA-N
SMILES: OC(CC(O)=O)C(O)=O.CN(C)CCC1=CNC2=CC=C(CS(=O)(=O)N3CCCC3)C=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results