accessibility menu, dialog, popup

UserName

H2NNH3NH2NNH3NOSOOO

CAS RN 1811-28-5

IUPAC Name: bis(6-aminoacridin-3-aminium) sulfate
Synonyms: bis(6-aminoacridin-3-aminium) sulfate
Molecular Weight (g/mol): 516.58
Molecular Formula: C26H24N6O4S
InChi Key: YADYXCVYLIKQJX-UHFFFAOYSA-N
SMILES: [O-]S([O-])(=O)=O.NC1=CC2=NC3=CC([NH3+])=CC=C3C=C2C=C1.NC1=CC2=NC3=CC([NH3+])=CC=C3C=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results