accessibility menu, dialog, popup

UserName

BrHNO

CAS RN 1810-66-8

IUPAC Name: 6-bromo-1,2-dihydroquinolin-2-one
Synonyms: 6-bromo-1H-quinolin-2-one
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: YLAFBGATSQRSTB-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)C=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results