CAS RN 180509-18-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(Alpha-D-Mannopyranosyl)-L-tryptophan, TRC
CAS: 180509-18-6 Molecular Formula: C17H22N2O7 Molecular Weight (g/mol): 366.37 Synonym: C2-α-D-Mannopyranosyltryptophan,2-α-D-Mannopyranosyl-L-tryptophan,2-α-D-Mannopyranosyl-L-tryptophan IUPAC Name: (S)-2-amino-3-(2-((2R,3S,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-1H-indol-3-yl)propanoic acid SMILES: N[C@@H](CC1=C([C@@H]2[C@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)NC3=C1C=CC=C3)C(O)=O