accessibility menu, dialog, popup

UserName

CH3SiH3CCH3OH2C

CAS RN 18042-43-8

IUPAC Name: trimethyl[2-(prop-2-en-1-yl)phenoxy]silane
Synonyms: trimethyl(2-(prop-2-en-1-yl)phenoxy)silane
Molecular Weight (g/mol): 206.36
Molecular Formula: C12H18OSi
InChi Key: LJIXCUHZKDSCQX-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=CC=CC=C1CC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results