accessibility menu, dialog, popup

UserName

SFFFFFFO

CAS RN 1800248-21-8

IUPAC Name: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 250.16
Molecular Formula: C7H4F6OS
InChi Key: FQOHJVDBMWSHSA-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results