CAS RN 1800102-50-4
CAS RN 1800102-50-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ropinirole Methylene Dimer, TRC
CAS: 1800102-50-4 Molecular Formula: C33H48N4O2 Molecular Weight (g/mol): 532.76 Synonym: 3,3'-Methylenebis(7-(2-(dipropylamino)ethyl)indolin-2-one) IUPAC Name: 4-[2-(dipropylamino)ethyl]-3-[[4-[2-(dipropylamino)ethyl]-2-oxo-1,3-dihydroindol-3-yl]methyl]-1,3-dihydroindol-2-one SMILES: CCCN(CCC)CCc1cccc2NC(=O)C(CC3C(=O)Nc4cccc(CCN(CCC)CCC)c34)c12