CAS RN 1797131-50-0
CAS RN 1797131-50-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Dechloro-4-(3-chlorophenyl) Haloperidol Decanoate, TRC
CAS: 1797131-50-0 Molecular Formula: C37 H45 Cl F N O3 Molecular Weight (g/mol): 606.21 Synonym: Haloperidol Decanoate EP Impurity F IUPAC Name: [4-[4-(3-chlorophenyl)phenyl]-1-[4-(4-fluorophenyl)-4-oxo-butyl]-4-piperidyl] decanoate SMILES: CCCCCCCCCC(=O)OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(cc3)c4cccc(Cl)c4