accessibility menu, dialog, popup

UserName

OHNNHNN

CAS RN 179057-20-6

IUPAC Name: [3-(2H-1,2,3,4-tetrazol-5-yl)phenyl]methanol
Synonyms: [3-(2H-1,2,3,4-tetrazol-5-yl)phenyl]methanol
Molecular Weight (g/mol): 176.18
Molecular Formula: C8H8N4O
InChi Key: YGESRLFHDANLIL-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results