accessibility menu, dialog, popup

UserName

OHOOHOOHONNEu(E/Z)(E/Z)(E/Z)

CAS RN 17904-83-5

IUPAC Name: 1,10-phenanthroline tris(3-hydroxy-1,3-diphenylprop-2-en-1-one) europium
Synonyms: tris(3-hydroxy-1,3-diphenylprop-2-en-1-one) phen europium
Molecular Weight (g/mol): 1004.95
Molecular Formula: C57H44EuN2O6
InChi Key: DYKOLWWJTALFFU-UHFFFAOYSA-N
SMILES: [Eu].C1=CN=C2C(C=CC3=CC=CN=C23)=C1.OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results