CAS RN 1783-84-2
CAS RN 1783-84-2
IUPAC Name:
(8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
Synonyms:
dihomo-gamma-linolenic acid gamma-homolinolenic acid 8c,11c,14c-eicosatrienoic acid 8, 11, 14-eicosatrienoic acid star gla (gnc) 8c,11c,14c-eicosatriensaeure homo-gamma-linolenic acid homo-gamma-linolensaeure 8,11,14-eicosatrienoic acid bishomo-gamma-linolenic acid
Molecular Weight (g/mol):
306.49
Molecular Formula:
C20H34O2
InChi Key:
HOBAELRKJCKHQD-QNEBEIHSSA-N
SMILES:
CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(O)=O
1
–
1
of
1
results
1
cis-8,11,14-Eicosatrienoic Acid, TRC
CAS: 1783-84-2 Molecular Formula: C20 H34 O2 Molecular Weight (g/mol): 306.48 Synonym: 8,11,14-Eicosatrienoic acid, (Z,Z,Z)- (8CI),(8Z,11Z,14Z)-8,11,14-Eicosatrienoic acid,(Z,Z,Z)-8,11,14-Eicosatrienoic acid,8,11,14-Eicosatrienoic acid,8,11,14-all-cis-Eicosatrienoic acid,8Z,11Z,14Z-Eicosatrienoic acid,Bishomo-γ-linolenic acid,DGLA,Dihomo-γ-linolenic acid,Diroleuton,Ro 12-1989,all-cis-8,11,14-Eicosatrienoic acid,cis-8,11,14-Eicosatrienoic acid,cis-8,cis-11,cis-14-Eicosatrienoic acid,γ-Homolinolenic acid IUPAC Name: (8Z,11Z,14Z)-icosa-8,11,14-trienoic acid SMILES: CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(=O)O