accessibility menu, dialog, popup

UserName

HBrBrON

CAS RN 17694-68-7

IUPAC Name: hydrogen 2-bromo-1-(pyridin-3-yl)ethan-1-one bromide
Synonyms: hydrogen 2-bromo-1-(pyridin-3-yl)ethanone bromide
Molecular Weight (g/mol): 280.95
Molecular Formula: C7H7Br2NO
InChi Key: WDTSYONULAZKIE-UHFFFAOYSA-N
SMILES: [H+].[Br-].BrCC(=O)C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)