accessibility menu, dialog, popup

UserName

CAS RN 175592-59-3

BSOHOOH

CAS RN 175592-59-3

IUPAC Name: (5-formylthiophen-3-yl)boronic acid
Synonyms: 5-formylthiophen-3-ylboronic acid
Molecular Weight (g/mol): 155.96
Molecular Formula: C5H5BO3S
InChi Key: ZCUFJAORCNFWOF-UHFFFAOYSA-N
SMILES: OB(O)C1=CSC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results