CAS RN 17533-08-3
IUPAC Name:
1,2,4-trifluoro-5-iodobenzene
Synonyms:
1,2,4-trifluoro-5-iodobenzene
Molecular Weight (g/mol):
257.98
Molecular Formula:
C6H2F3I
InChi Key:
UPCKPWDYXMTASO-UHFFFAOYSA-N
SMILES:
FC1=CC(F)=C(I)C=C1F
1
–
1
of
1
results
1
2,4,5-Trifluoro-1-iodobenzene, 95%, Thermo Scientific™
CAS: 17533-08-3 Molecular Formula: C6H2F3I Molecular Weight (g/mol): 257.98 MDL Number: MFCD04973898 InChI Key: UPCKPWDYXMTASO-UHFFFAOYSA-N Synonym: 2,4,5-trifluoroiodobenzene,1-iodo-2,4,5-trifluorobenzene,acmc-209e9x PubChem CID: 2760713 IUPAC Name: 1,2,4-trifluoro-5-iodobenzene SMILES: FC1=CC(F)=C(I)C=C1F