accessibility menu, dialog, popup

UserName

CH3H3COCl

CAS RN 17530-69-7

IUPAC Name: 3-chloro-5,5-dimethylcyclohex-2-en-1-one
Synonyms: 3-chloro-5,5-dimethylcyclohex-2-en-1-one
Molecular Weight (g/mol): 158.63
Molecular Formula: C8H11ClO
InChi Key: PJYTYJGMJDIKEJ-UHFFFAOYSA-N
SMILES: CC1(C)CC(Cl)=CC(=O)C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results