accessibility menu, dialog, popup

UserName

CH3H3CCH3NNOCl

CAS RN 175205-41-1

IUPAC Name: 5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole
Synonyms: 5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 174.63
Molecular Formula: C7H11ClN2O
InChi Key: AIVSLDAXUIRICJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=NC(CCl)=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results