CAS RN 175205-36-4
IUPAC Name:
(2-aminoethyl)amino 4-methylbenzene-1-sulfonate
Synonyms:
(2-aminoethyl)amino 4-methylbenzenesulfonate
Molecular Weight (g/mol):
230.28
Molecular Formula:
C9H14N2O3S
InChi Key:
RWUMUZOGZPNCIL-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)ONCCN
1
–
1
of
1
results
1
N-[[(4-Methylphenyl)sulfonyl]oxy]ethane -1,2-diamine, ≥95%, Thermo Scientific™
CAS: 175205-36-4 Molecular Formula: C9H14N2O3S Molecular Weight (g/mol): 230.282 MDL Number: MFCD00276588 InChI Key: RWUMUZOGZPNCIL-UHFFFAOYSA-N Synonym: 2-tosyloxy amino ethanamine,2-aminoethyl amino 4-methylbenzenesulfonate,benzenesulfonic acid,4-methyl-, 2-aminoethyl azanyl ester,n1-4-methylphenyl sulfonyl oxy ethane-1,2-diamine,2-aminoethylamino 4-methylbenzenesulfonate,2-4-methylphenyl sulfonyl-oxy amino ethanamine,n∼1∼-4-methylbenzene-1-sulfonyl oxy ethane-1,2-diamine,n∼1∼-4-methylphenyl sulfonyl oxy ethane-1,2-diamine PubChem CID: 2796248 IUPAC Name: (2-aminoethylamino) 4-methylbenzenesulfonate SMILES: CC1=CC=C(C=C1)S(=O)(=O)ONCCN