accessibility menu, dialog, popup

UserName

CAS RN 175204-80-5

NFFFH2N

CAS RN 175204-80-5

IUPAC Name: 4-(trifluoromethyl)pyridin-3-amine
Synonyms: 4-(trifluoromethyl)pyridin-3-amine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: DROFUWWORPKKSI-UHFFFAOYSA-N
SMILES: NC1=C(C=CN=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results