accessibility menu, dialog, popup

UserName

CAS RN 175203-84-6

IUPAC Name: 5-(trifluoromethoxy)-1H-indole-2-carboxylic acid
Synonyms: 5-(trifluoromethoxy)-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 245.16
Molecular Formula: C10H6F3NO3
InChi Key: DJJJSHFPEISHFN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(OC(F)(F)F)=CC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)