accessibility menu, dialog, popup

UserName

CAS RN 175201-94-2

H3CNNNClOOHH3C

CAS RN 175201-94-2

IUPAC Name: 4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid
Synonyms: 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylic acid
Molecular Weight (g/mol): 225.63
Molecular Formula: C9H8ClN3O2
InChi Key: LHEUUYZQYKWDSY-UHFFFAOYSA-N
SMILES: CN1N=C(C)C2=C(Cl)C(=CN=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results