accessibility menu, dialog, popup

UserName

CH3H3CSOOSH2NOOH3C

CAS RN 175201-72-6

IUPAC Name: methyl 3-amino-4-(propane-2-sulfonyl)thiophene-2-carboxylate
Synonyms: methyl 3-amino-4-(propane-2-sulfonyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 263.33
Molecular Formula: C9H13NO4S2
InChi Key: LSLWECVRXQXIRH-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(=CS1)S(=O)(=O)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results