accessibility menu, dialog, popup

UserName

CH3ONSOOH3CH3CCH3(E/Z)

CAS RN 175201-67-9

IUPAC Name: 3-ethoxy-2-(2-methylpropane-2-sulfonyl)prop-2-enenitrile
Synonyms: 3-ethoxy-2-(2-methylpropane-2-sulfonyl)prop-2-enenitrile
Molecular Weight (g/mol): 217.28
Molecular Formula: C9H15NO3S
InChi Key: UEZQDEIGENVNHG-UHFFFAOYSA-N
SMILES: CCOC=C(C#N)S(=O)(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)