CAS RN 17508-17-7
CAS RN 17508-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
O-(2,4-Dinitrophenyl)hydroxylamine, TRC
CAS: 17508-17-7 Molecular Formula: C6H5N3O5 Molecular Weight (g/mol): 199.12 Synonym: 1-Aminooxy-2,4-dinitrobenzene,2,4-Dinitrophenoxyamine,2,4-Nitrophenoxyamine,DNPA,NSC 148499; SMILES: C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])ON