CAS RN 174913-09-8
CAS RN 174913-09-8
IUPAC Name:
1-bromo-4-chloro-2-methoxybenzene
Synonyms:
1-bromo-4-chloro-2-methoxybenzene
Molecular Weight (g/mol):
221.48
Molecular Formula:
C7H6BrClO
InChi Key:
CQGYLDZGJLVLMK-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=CC(Cl)=C1
1
–
1
of
1
results
1
2-Bromo-5-chloroanisole, 98+%
CAS: 174913-09-8 Molecular Formula: C7H6BrClO Molecular Weight (g/mol): 221.478 MDL Number: MFCD03790889 InChI Key: CQGYLDZGJLVLMK-UHFFFAOYSA-N Synonym: 2-bromo-5-chloroanisole,benzene, 1-bromo-4-chloro-2-methoxy,pubchem5265,acmc-1c20c,ksc497i0p,2-bromo-5-chloroanisole, 98+%,2-methoxy-4-chloro-1-bromobenzene,1-bromo-4-chloro-2-methoxy-benzene PubChem CID: 17984845 IUPAC Name: 1-bromo-4-chloro-2-methoxybenzene SMILES: COC1=C(C=CC(=C1)Cl)Br