accessibility menu, dialog, popup

UserName

OOO

CAS RN 17422-74-1

IUPAC Name: 4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 174.16
Molecular Formula: C10H6O3
InChi Key: FSMYWBQIMDSGQP-UHFFFAOYSA-N
SMILES: O=CC1=COC2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)