accessibility menu, dialog, popup

UserName

CAS RN 1737-68-4

IUPAC Name: 4-fluoro-2,3-dimethylaniline
Synonyms: 4-fluoro-2,3-dimethylaniline
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: LKMJLYCMLAPNDB-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(F)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results