accessibility menu, dialog, popup

UserName

CAS RN 17249-82-0

IUPAC Name: 2-chloro-5-methylthiophene
Synonyms: 2-chloro-5-methylthiophene
Molecular Weight (g/mol): 132.61
Molecular Formula: C5H5ClS
InChi Key: JSMMZMYGEVUURX-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)S1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results