accessibility menu, dialog, popup

UserName

H3CONHNNNNOOPOOOOOH3CNa

CAS RN 16980-89-5

IUPAC Name: sodium 6-(6-butanamido-9H-purin-9-yl)-7-(butanoyloxy)-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonyms: sodium 6-(6-butanamidopurin-9-yl)-7-(butanoyloxy)-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Weight (g/mol): 491.37
Molecular Formula: C18H23N5NaO8P
InChi Key: KRBZRVBLIUDQNG-UHFFFAOYNA-M
SMILES: [Na+].CCCC(=O)NC1=C2N=CN(C3OC4COP([O-])(=O)OC4C3OC(=O)CCC)C2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)