accessibility menu, dialog, popup

UserName

CAS RN 16919-58-7

NH4NH4PtClClClClClCl

CAS RN 16919-58-7

IUPAC Name: diammonium hexachloroplatinumdiuide
Synonyms: ammonium hexachloroplatinate ammonium platinum chloride ammonium platinic chloride ammonium chloroplatinate diammonium platinum hexachloride
Molecular Weight (g/mol): 443.86
Molecular Formula: Cl6H8N2Pt
InChi Key: PCCGQTHFYHJATL-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results