accessibility menu, dialog, popup

UserName

CAS RN 1686-22-2

CH3OHVOHH3COHCH3O

CAS RN 1686-22-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 205.15
Molecular Formula: C6H18O4V
InChi Key: IDIDIJSLBFQEKY-UHFFFAOYSA-N
SMILES: O=[V].CCO.CCO.CCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results