CAS RN 168479-96-7
CAS RN 168479-96-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-(2,4-Difluorophenyl)-2-(4H-1,2,4-triazol-4-yl)ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(2,4-Difluorophenyl)-2-(4H-1,2,4-triazol-4-yl)ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-(2,4-Difluorophenyl)-2-(4H-1,2,4-triazol-4-yl)ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.