accessibility menu, dialog, popup

UserName

CAS RN 1678-93-9

IUPAC Name: butylcyclohexane
Synonyms: butylcyclohexane
Molecular Weight (g/mol): 140.27
Molecular Formula: C10H20
InChi Key: GGBJHURWWWLEQH-UHFFFAOYSA-N
SMILES: CCCCC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results