accessibility menu, dialog, popup

UserName

CAS RN 16738-20-8

CH3NHNH2SH3C

CAS RN 16738-20-8

IUPAC Name: (2,4-dimethylphenyl)thiourea
Synonyms: 2,4-dimethylphenylthiourea
Molecular Weight (g/mol): 180.27
Molecular Formula: C9H12N2S
InChi Key: KAGLPYRXTCQWHU-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(N)=S)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results