accessibility menu, dialog, popup

UserName

H3COOOOH

CAS RN 16695-14-0

IUPAC Name: 3-methoxy-3-oxopropanoic acid
Synonyms: 3-methoxy-3-oxopropanoic acid
Molecular Weight (g/mol): 118.09
Molecular Formula: C4H6O4
InChi Key: PBVZQAXFSQKDKK-UHFFFAOYSA-N
SMILES: COC(=O)CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results