accessibility menu, dialog, popup

UserName

BrNHN

CAS RN 16681-56-4

IUPAC Name: 2-bromo-1H-imidazole
Synonyms: 2-bromo-1H-imidazole
Molecular Weight (g/mol): 146.98
Molecular Formula: C3H3BrN2
InChi Key: AXHRGVJWDJDYPO-UHFFFAOYSA-N
SMILES: BrC1=NC=CN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)