accessibility menu, dialog, popup

UserName

ClSiCl

CAS RN 1631-84-1

IUPAC Name: dichloro(phenyl)silyl
Synonyms: dichloro(phenyl)silyl
Molecular Weight (g/mol): 176.09
Molecular Formula: C6H5Cl2Si
InChi Key: XNAFLNBULDHNJS-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)