CAS RN 162744-59-4
CAS RN 162744-59-4
IUPAC Name:
(4-bromo-2,6-difluorophenyl)methanol
Synonyms:
(4-bromo-2,6-difluorophenyl)methanol
Molecular Weight (g/mol):
223.02
Molecular Formula:
C7H5BrF2O
InChi Key:
LSRHFWSNUFIKER-UHFFFAOYSA-N
SMILES:
OCC1=C(F)C=C(Br)C=C1F
1
–
1
of
1
results
1
4-Bromo-2,6-difluorobenzyl alcohol, 97%
CAS: 162744-59-4 Molecular Formula: C7H5BrF2O Molecular Weight (g/mol): 223.017 MDL Number: MFCD03094461 InChI Key: LSRHFWSNUFIKER-UHFFFAOYSA-N Synonym: 4-bromo-2,6-difluorobenzyl alcohol,4-bromo-2,6-difluorophenyl methanol,4-bromo-2,6-difluorobenzylalcohol,benzenemethanol, 4-bromo-2,6-difluoro,4-bromo-2,6-difluoro-benzenemethanol,2,6-difluoro-4-bromobenzyl alcohol,4-bromo-2,6-difluoro-phenyl methanol,2,6-difluoro-4-bromo benzyl alcohol,pubchem4882,acmc-209dox PubChem CID: 2773303 IUPAC Name: (4-bromo-2,6-difluorophenyl)methanol SMILES: C1=C(C=C(C(=C1F)CO)F)Br