accessibility menu, dialog, popup

UserName

OONH2

CAS RN 16081-45-1

IUPAC Name: 2,3-dihydro-1,4-benzodioxin-5-amine
Synonyms: 2,3-dihydro-1,4-benzodioxin-5-amine
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: DMLRSJNZORFCBD-UHFFFAOYSA-N
SMILES: NC1=C2OCCOC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results