accessibility menu, dialog, popup

UserName

NHNONHHO

CAS RN 160807-49-8

IUPAC Name: 3-(hydroxyamino)-1H,2'H-[2,3'-biindol]-2'-one
Synonyms: 3-(hydroxyamino)-1H-[2,3'-biindol]-2'-one
Molecular Weight (g/mol): 277.28
Molecular Formula: C16H11N3O2
InChi Key: LDEWQRSYYHTQRA-UHFFFAOYSA-N
SMILES: ONC1=C(NC2=CC=CC=C12)C1=C2C=CC=CC2=NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results