accessibility menu, dialog, popup

UserName

CAS RN 1608-26-0

H3CNCH3PNCH3CH3NCH3H3C

CAS RN 1608-26-0

IUPAC Name: [bis(dimethylamino)phosphanyl]dimethylamine
Synonyms: [bis(dimethylamino)phosphanyl]dimethylamine
Molecular Weight (g/mol): 163.21
Molecular Formula: C6H18N3P
InChi Key: XVDBWWRIXBMVJV-UHFFFAOYSA-N
SMILES: CN(C)P(N(C)C)N(C)C
Boiling Point 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)