accessibility menu, dialog, popup

UserName

CAS RN 160388-55-6

IUPAC Name: {4-[(1H-pyrazol-1-yl)methyl]phenyl}methanol
Synonyms: [4-(pyrazol-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: ADLULGOPZPFWPA-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CN2C=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results