CAS RN 160369-85-7
CAS RN 160369-85-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Bromo-4-chloro-3-indolyl-Alpha-D-N-acetylneuraminic Acid, Sodium Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.