accessibility menu, dialog, popup

UserName

CAS RN 16013-85-7

ONOClNCl

CAS RN 16013-85-7

IUPAC Name: 2,6-dichloro-3-nitropyridine
Synonyms: 2,6-dichloro-3-nitropyridine
Molecular Weight (g/mol): 192.98
Molecular Formula: C5H2Cl2N2O2
InChi Key: SHCWQWRTKPNTEM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)N=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)