CAS RN 1592-00-3
IUPAC Name:
1-bromo-2-isocyanatobenzene
Synonyms:
1-bromo-2-isocyanatobenzene
Molecular Weight (g/mol):
198.02
Molecular Formula:
C7H4BrNO
InChi Key:
GOOVAYJIVMBWPP-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC=C1N=C=O
1
–
1
of
1
results
1
2-Bromophenyl isocyanate, 97%
CAS: 1592-00-3 Molecular Formula: C7H4BrNO Molecular Weight (g/mol): 198.019 MDL Number: MFCD00001995 InChI Key: GOOVAYJIVMBWPP-UHFFFAOYSA-N Synonym: 2-bromophenyl isocyanate,2-bromophenylisocyanate,benzene, 1-bromo-2-isocyanato,isocyanic acid 2-bromophenyl ester,2-bromobenzenisocyanate,2bromophenylisocyanate,2-bromophenylisocynate,o-bromophenyl isocyanate,acmc-209djl,2-bromo phenylisocyanate PubChem CID: 137097 IUPAC Name: 1-bromo-2-isocyanatobenzene SMILES: C1=CC=C(C(=C1)N=C=O)Br