accessibility menu, dialog, popup

UserName

CAS RN 1591-99-7

IUPAC Name: 1-isocyanato-2,3-dimethylbenzene
Synonyms: 2,3-dimethylphenyl isocyanate
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: KNHJIEOCVVIBIV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(N=C=O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results