accessibility menu, dialog, popup

UserName

CAS RN 1586-73-8

H3CSiH3CCH3NSiCH3CH3H3CSiCH3CH3H3C

CAS RN 1586-73-8

IUPAC Name: tris(trimethylsilyl)amine
Synonyms: tris(trimethylsilyl)amine
Molecular Weight (g/mol): 233.58
Molecular Formula: C9H27NSi3
InChi Key: PEGHITPVRNZWSI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N([Si](C)(C)C)[Si](C)(C)C
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)