accessibility menu, dialog, popup

UserName

CH3NBrN

CAS RN 15803-02-8

IUPAC Name: 4-bromo-1-methyl-1H-pyrazole
Synonyms: 4-bromo-1-methylpyrazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: IXJSDKIJPVSPKF-UHFFFAOYSA-N
SMILES: CN1C=C(Br)C=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)