CAS RN 1565-80-6
IUPAC Name:
2-methylbutan-1-ol
Synonyms:
2-methyl-1-butanol (1)-2-methylbutan-1-ol active amyl alcohol primary active amyl alcohol active primary amyl alcohol methyl-2-butan-1-ol
Molecular Weight (g/mol):
88.15
Molecular Formula:
C5H12O
InChi Key:
QPRQEDXDYOZYLA-UHFFFAOYNA-N
SMILES:
CCC(C)CO
1
–
1
of
1
results
1
(S)-(-)-2-Methylbutanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.