CAS RN 15627-86-8
CAS RN 15627-86-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
No Refinements
1
–
0
of
0
results
1
Calcium perchlorate tetrahydrate, pure
CAS: 15627-86-8 Molecular Formula: CaCl2O8·4H2O Molecular Weight (g/mol): 311.04 MDL Number: MFCD00191895