accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3CClIClClCl

CAS RN 15625-59-9

IUPAC Name: tetrabutylazanium; tetrachloro-λ³-iodanuide
Synonyms: tetrabutylammonium ion; tetrachloro-λ³-iodanuide
Molecular Weight (g/mol): 511.18
Molecular Formula: C16H36Cl4IN
InChi Key: HBZJAVINJZUOEH-UHFFFAOYSA-N
SMILES: Cl[I-](Cl)(Cl)Cl.CCCC[N+](CCCC)(CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results